Orders LinkContactUsLinkComponent
Portable Air Compressor, Electron Microscopy Sciences
Portable Air Compressor, Electron Microscopy Sciences
Portable Air Compressor
Catalog # 101412-916
Supplier #:  64195
Portable Air Compressor, Electron Microscopy Sciences
Catalog # 101412-916
Portable Air Compressor, Electron Microscopy Sciences
Portable Air Compressor, Electron Microscopy Sciences
Portable Air Compressor
Price
$265.98
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pressurize the Sure Shot® Sprayers with this rechargeable battery powered Air Compressor.
The Compressor generates up to 220 PSI, the battery recharge unit by the AC 115V adapter or DC 12V adapter included. Completely portable, no trailing cords.
Specifications
  • Description:
    Portable Air Compressor