Extra Long Transfer Pipettes, Electron Microscopy Sciences
About this item
These transfer pipettes are ideal for subculturing, harvesting, sampling 15, 50, 100 ml tubes and flasks, removing supernatant and media from cell cultures, wash cells, post centrifugation to remove cell layers, layer or transfers in columns, loading gels, field testing, and blood culture bottle. These pipettes come sterile and non-sterile. Sterile pipettes are individually wrapped , then are bulk packaged.
4 Options Available Below
Product Details & Documents
These transfer pipettes are ideal for subculturing, harvesting, sampling 15, 50, 100 ml tubes and flasks, removing supernatant and media from cell cultures, wash cells, post centrifugation to remove cell layers, layer or transfers in columns, loading gels, field testing, and blood culture bottle. These pipettes come sterile and non-sterile. Sterile pipettes are individually wrapped , then are bulk packaged.
Product Family Options
Product Information
- VolumeØ×LSterilityDrop sizeBulb drawAvailabilityPrice
- Supplier Number: 70968-26
- Supplier Number: 70968-27
- Supplier Number: 70968-26S
- Supplier Number: 70968-27S