Order Entry
Puerto Rico
ContactUsLinkComponent
Locking Cart, Electron Microscopy Sciences
Locking Cart, Electron Microscopy Sciences
  102095-204
undefined
Locking Cart, Electron Microscopy Sciences
  102095-204
 :  64382-50

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Non-Ferrous Locking Roll-Top Cart
  • Cat. No.:
    102095-204
  • Supplier no.:
    64382-50

 

 

This locking with roll-top cart has six drawers, two keys with roll top tambour door.

  • HDPE durable top
  • Smooth rolling on heavy-duty with two lockable casters
  • Door lockable for security
  • Two keys provided

Measures: 12” (W) x 36” (H) x 19” (D)