Orders LinkContactUsLinkComponent
INFINITY 3-1 Cooled CCD Cameras, Electron Microscopy Sciences
INFINITY 3-1 Cooled CCD Cameras, Electron Microscopy Sciences
INFINITY 3-1 Cooled CCD Cameras, Electron Microscopy Sciences
Certificates
INFINITY 3-1 Cooled CCD Cameras, Electron Microscopy Sciences
INFINITY 3-1 Cooled CCD Cameras, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For low light fluorescence applications, the INFINITY CCD USB 2.0 cameras offer optimal color reproduction and extreme sensitivity.
  • 1 megapixel resolution
  • High signal to noise ratio for low light, long exposure applications
  • Available with cooling to 25° C below ambient
  • Fast frame rates
  • Low noise electronics
  • Available with and without cooling
The Sony ICX285 ExView HAD sensor has very a high dynamic range, excellent sensitivity and a 12-bit digital output. Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest. Cooling to 25° C below ambient is available for low-light conditions and where high dynamic range is required.
Applications: Brightfield, Darkfield, DIC, Live Cell Imaging, Histology, Pathology, Cytology, Defect Analysis, Semiconductor Inspection, Metrology, Low Light Fluorescence, Chemiluminescence, Bioluminescence, Flow Analysis, GFP, FISH, NIR, FRET.
Specifications for Product Family
Specifications
  • Catalog No:
  • 102091-460
  • 102091-462
  • 102091-474
  • 102091-476
  • Model:
  • INFINITY3-1C
  • INFINITY3-1M
  • INFINITY3-1U
  • INFINITY3-1U
  • Sensor:
  • Cooled CCD Color
  • Cooled CCD Mono
  • CCD Color
  • CCD Mono
  • Sensor size:
  • 2/3"
  • 2/3"
  • 2/3"
  • 2/3"
  • Frame Rate:
  • 15 fps
  • 15 fps
  • 30 fps
  • 30 fps
  • Interfaces:
  • USB 2.0
  • USB 2.0
  • USB 2.0
  • USB 2.0
Specifications
Product Family Options
Product Information
  • Resolution
    Sensor type
    Availability
    Price
  • 102091-460
    INFINITY3-1C Cooled Color Camera
    INFINITY3-1C Cooled Color Camera
    1.4 MP
    CCD
    Log in for availability
    Log in for availability
    Each
    Each - $14,496.08
    $14,496.08
    Each
       $14,496.08
    Supplier Number: 95110
    Specifications:
    More
    Specifications
    Cat. No.
    102091-460
    Model
    INFINITY3-1C
    Sensor
    Cooled CCD Color
    Sensor size
    2/3"
    Frame Rate
    15 fps
    Interfaces
    USB 2.0
  • 102091-462
    INFINITY3-1M Cooled Monochrome Camera
    INFINITY3-1M Cooled Monochrome Camera
    1.4 MP
    CCD
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    102091-462
    Model
    INFINITY3-1M
    Sensor
    Cooled CCD Mono
    Sensor size
    2/3"
    Frame Rate
    15 fps
    Interfaces
    USB 2.0
  • 102091-474
    INFINITY3-1U Color Camera
    INFINITY3-1U Color Camera
    1.4 MP
    CCD
    Discontinued
    Supplier Number: 95119
    Specifications:
    More
    Specifications
    Cat. No.
    102091-474
    Model
    INFINITY3-1U
    Sensor
    CCD Color
    Sensor size
    2/3"
    Frame Rate
    30 fps
    Interfaces
    USB 2.0
  • 102091-476
    INFINITY3-1U Monochrome Camera
    INFINITY3-1U Monochrome Camera
    1.4 MP
    CCD
    Log in for availability
    Log in for availability
    Each
    Each - $9,478.23
    $9,478.23
    Each
       $9,478.23
    Supplier Number: 95120
    Specifications:
    More
    Specifications
    Cat. No.
    102091-476
    Model
    INFINITY3-1U
    Sensor
    CCD Mono
    Sensor size
    2/3"
    Frame Rate
    30 fps
    Interfaces
    USB 2.0