INFINITY 1 CMOS Cameras, Electron Microscopy Sciences
Lumenera’s INFINITY1 CMOS USB 2.0 digital cameras are designed to be a cost-effective, versatile solution for clinical, life science, materials science and educational professionals.
- 2, 3 and 5 megapixel resolution
- Perfect for documentation and archiving applications
- Fast frame rates
- Auto white balance
- Full exposure control
- Programmable gain
- Sub-windowing and region of interest
- An excellent fit for documentation and archiving applications
The camera is availabe with 1.3 to 5 megapixel resolution.The INFINITY1-2C is the newest camera in the series and features a unique color sensor with 1600 x 1200 resolution and on-board processing. This camera delivers outstanding image quality and excellent value.
Applications: Brightfield, Darkfield, DIC, Live Cell Imaging, Histology, Pathology, Cytology, Defect Analysis, Semiconductor Inspection, and Metrology.
Lumenera’s INFINITY1 CMOS USB 2.0 digital cameras are designed to be a cost-effective, versatile solution for clinical, life science, materials science and educational professionals.
- 2, 3 and 5 megapixel resolution
- Perfect for documentation and archiving applications
- Fast frame rates
- Auto white balance
- Full exposure control
- Programmable gain
- Sub-windowing and region of interest
- An excellent fit for documentation and archiving applications
The camera is availabe with 1.3 to 5 megapixel resolution.The INFINITY1-2C is the newest camera in the series and features a unique color sensor with 1600 x 1200 resolution and on-board processing. This camera delivers outstanding image quality and excellent value.
Applications: Brightfield, Darkfield, DIC, Live Cell Imaging, Histology, Pathology, Cytology, Defect Analysis, Semiconductor Inspection, and Metrology.
- Catalog No:
- 102091-442
- 102091-444
- 102091-446
- 102091-448
- Model:
- INFINITY1-2C
- INFINITY1-1M
- INFINITY1-3C
- INFINITY1-5C
- Sensor:
- CMOS Color
- CMOS Mono
- CMOS Color
- CMOS Color
- Sensor size:
- 1/2"
- 1/2"
- 1/2"
- 1/2.5"
- Pixels (W×H):
- 1600×1200
- 1280×1024
- 2048×1536
- 2592×1944
- Frame Rate:
- 15 fps
- 30 fps
- 12 fps
- 7 fps
- Interfaces:
- USB 2.0
- USB 2.0
- USB 2.0
- USB 2.0
Product Family Options
- DescriptionResolutionSensor type
- : 95000
Specifications
Cat. No.102091-442ModelINFINITY1-2CSensorCMOS ColorSensor size1/2"Pixels (W×H)1600×1200Frame Rate15 fpsInterfacesUSB 2.0 - : 95100
Specifications
Cat. No.102091-444ModelINFINITY1-1MSensorCMOS MonoSensor size1/2"Pixels (W×H)1280×1024Frame Rate30 fpsInterfacesUSB 2.0 - : 95102
Specifications
Cat. No.102091-446ModelINFINITY1-3CSensorCMOS ColorSensor size1/2"Pixels (W×H)2048×1536Frame Rate12 fpsInterfacesUSB 2.0 - : 95103
Specifications
Cat. No.102091-448ModelINFINITY1-5CSensorCMOS ColorSensor size1/2.5"Pixels (W×H)2592×1944Frame Rate7 fpsInterfacesUSB 2.0