Orders LinkContactUsLinkComponent
INFINITY 2-1 C CCD Color Camera, Electron Microscopy Sciences
INFINITY 2-1 C CCD Color Camera, Electron Microscopy Sciences
INFINITY 2-1 C CCD Color Camera, Electron Microscopy Sciences
INFINITY 2-1 C CCD Color Camera, Electron Microscopy Sciences
INFINITY 2-1 C CCD Color Camera, Electron Microscopy Sciences
Equipped with a high-quality, Sony CCD sensor, INFINITY CCD USB 2.0 cameras offer excellent sensitivity, high dynamic range and a 12-bit digital output.
  • 1, 2 and 3 megapixel resolution
  • Higher dynamic range for quantitative analysis
  • Fast frame rates
  • Low noise electronics
Features include binning, auto white balance, full exposure control, programmable gain, sub-windowing and region of interest. It is ideal for higherend scientific, medical, ophthalmic and life science applications.
Applications: Brightfield, Darkfield, DIC, Live Cell Imaging, Histology, Pathology, Cytology, Defect Analysis, Semiconductor Inspection, Metrology, Gel Documentation, and Moderate Light Fluorescence.
  • Catalog No:
  • 102091-450
  • 102091-452
  • 102091-454
  • 102091-456
  • 102091-458
  • 102091-472
  • 102091-478
  • Model:
  • INFINITY2-1C
  • INFINITY2-1M
  • INFINITY2-2C
  • INFINITY2-2M
  • INFINITY2-3C
  • INFINITY2-5C
  • INFINITY2-5M
  • Sensor:
  • CCD Color
  • CCD Mono
  • CCD Color
  • CCD Mono
  • CCD Color
  • CCD Color
  • CCD Mono
  • Sensor size:
  • 1/2"
  • 1/2"
  • 1/1.8"
  • 1/1.8"
  • 1/1.8"
  • 2/3"
  • 2/3"
  • Frame Rate:
  • 30 fps
  • 30 fps
  • 12 fps
  • 12 fps
  • 5 fps
  • 9 fps
  • 9 fps
  • Interfaces:
  • USB 2.0
  • USB 2.0
  • USB 2.0
  • USB 2.0
  • USB 2.0
  • USB 2.0
  • USB 2.0
Product Family Options
  • Description
    Resolution
    Sensor type
  • INFINITY2-1C Color Camera
    1.4 MP
    CCD
    Each
    Each - $5,217.36
    $5,217.36
    Each
       $5,217.36
     : 95105
    Specifications
    Cat. No.
    102091-450
    Model
    INFINITY2-1C
    Sensor
    CCD Color
    Sensor size
    1/2"
    Frame Rate
    30 fps
    Interfaces
    USB 2.0
  • INFINITY2-1M Monochrome Camera
    1.4 MP
    CCD
    Each
    Each - $5,217.36
    $5,217.36
    Each
       $5,217.36
     : 95106
    Specifications
    Cat. No.
    102091-452
    Model
    INFINITY2-1M
    Sensor
    CCD Mono
    Sensor size
    1/2"
    Frame Rate
    30 fps
    Interfaces
    USB 2.0
  • INFINITY2-2C Color Camera
    2 MP
    CCD
    Each
    Each - $8,831.70
    $8,831.70
    Each
       $8,831.70
     : 95107
    Specifications
    Cat. No.
    102091-454
    Model
    INFINITY2-2C
    Sensor
    CCD Color
    Sensor size
    1/1.8"
    Frame Rate
    12 fps
    Interfaces
    USB 2.0
  • INFINITY2-2M Monochrome Camera
    2 MP
    CCD
    Each
    Each - $7,734.44
    $7,734.44
    Each
       $7,734.44
     : 95108
    Specifications
    Cat. No.
    102091-456
    Model
    INFINITY2-2M
    Sensor
    CCD Mono
    Sensor size
    1/1.8"
    Frame Rate
    12 fps
    Interfaces
    USB 2.0
  • INFINITY2-3C Color Camera
    3.3 MP
    CCD
    Each
    Each - $8,028.42
    $8,028.42
    Each
       $8,028.42
     : 95109
    Specifications
    Cat. No.
    102091-458
    Model
    INFINITY2-3C
    Sensor
    CCD Color
    Sensor size
    1/1.8"
    Frame Rate
    5 fps
    Interfaces
    USB 2.0
  • INFINITY2-5C Color Camera
    5 MP
    CCD
     : 95118
    Specifications
    Cat. No.
    102091-472
    Model
    INFINITY2-5C
    Sensor
    CCD Color
    Sensor size
    2/3"
    Frame Rate
    9 fps
    Interfaces
    USB 2.0
  • INFINITY2-5M Monochrome Camera
    5 MP
    CCD
    Each
    Each - $10,035.75
    $10,035.75
    Each
       $10,035.75
     : 95121
    Specifications
    Cat. No.
    102091-478
    Model
    INFINITY2-5M
    Sensor
    CCD Mono
    Sensor size
    2/3"
    Frame Rate
    9 fps
    Interfaces
    USB 2.0