Non-ionic, low foaming phosphate free powdered enzyme cleaners for use in labware washers, manual and ultrasonic cleaning.
- Passes inhibitor residue test
- Dilute 1:200 and it has a pH 11.5
Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porselain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues.