About this item
Low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high performance removal of metal oxides, scale, salts and inorganic residues in lab-ware washers, parts washers, power-spray systems and manual and ultrasonic cleaning.
- Ideal for acid washing and rinsing
- Dilute 2:100 and it has a pH 2.5
Corrosion inhibited and free rinsing leaving no interfering residues.