- Description:UltraBed Low Viscosity Kit
- Size:2 x 100 ml
- Cat. No.:101410-644
- Supplier no.:14310
The kit consists of two components, equal amount, very low viscosity, (~65 cps), in convenient dispenser top bottles. To prepare the embedding resin, just mix equal amounts of the two components. The resin readily infiltrates into specimen and polymerizes to a clear, hard block overnight at 60°C.
- A Newly Introduced Low Viscosity Epoxy Embedding Kit
- Mixed resin can be used for both infiltration and embedding
- All components are shipped together as Non- Hazardous substances