Ilford Multigrade RC Warmtone Paper, Electron Microscopy Sciences
About this item
This is a direct replacement for Kodak Polycontrast IV RC B&W Papers. MULTIGRADE RC Warmtone Paper has the standard weight of (190g/m2) with a resin coated base. It is equally suitable for printing conventional black and white and XP2 SUPER negatives.
- Rich, Warm Blacks; Creamy Whites
- Excellent Toning Characteristics
- Developer: MULTIGRADE, or BROMOPHEN, or PQ UNIVERSAL
- Stop Bath : ILFOSTOP, or ILFOSTOP PRO
- Fixer: ILFORD RAPID FIXER or HYPAM
- For ILFORD processor: ILFORD 2000RT developer/replenisher & fixer/replenisher are recommended
- Glossy Surface
2 Options Available Below
Product Details & Documents
This is a direct replacement for Kodak Polycontrast IV RC B&W Papers. MULTIGRADE RC Warmtone Paper has the standard weight of (190g/m2) with a resin coated base. It is equally suitable for printing conventional black and white and XP2 SUPER negatives.
- Rich, Warm Blacks; Creamy Whites
- Excellent Toning Characteristics
- Developer: MULTIGRADE, or BROMOPHEN, or PQ UNIVERSAL
- Stop Bath : ILFOSTOP, or ILFOSTOP PRO
- Fixer: ILFORD RAPID FIXER or HYPAM
- For ILFORD processor: ILFORD 2000RT developer/replenisher & fixer/replenisher are recommended
- Glossy Surface
Product Family Options
Product Information
- SizeAvailabilityPrice
- 101381-632Multigrade RC Warmtone PaperSupplier Number: 83027-25
- 101381-630Multigrade RC Warmtone PaperSupplier Number: 83027-1C