Specifications
- Description:Lab Jacks
- Cat. No.:100498-354
- Supplier no.:63225-24
Specifications
About this item
This LabJack provides stable height adjustment and support for items in the lab such as flasks, baths or hot plate stirrers.
Top and bottom plates are constructed of anodized aluminum while the support mechanism is made of chrome plated steel.