ContactUsLinkComponent
Paterson Print Tongs, Electron Microscopy Sciences
Paterson Print Tongs, Electron Microscopy Sciences
  100492-250
Minority or Woman-Owned Business Enterprise
undefined
Paterson Print Tongs, Electron Microscopy Sciences
  100492-250
 :  74817
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Paterson Print Tong
  • Cat. No.:
    100492-250
  • Supplier no.:
    74817
These tongs have textured side panels which offer a firm grip and deeply notched “shoulders” which avoid slippage into the tray.
The tongs are all plastic and chemically resistant. Tongs are color-coded in light and dark gray to help minimize the risk of cross-contamination.