Orders LinkContactUsLinkComponent
Chessboard Squares Graticules, Electron Microscopy Sciences
Chessboard Squares Graticules, Electron Microscopy Sciences
Certificates
Chessboard Squares Graticules, Electron Microscopy Sciences
Chessboard Squares Graticules, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The dark squares are used as an alternative to simple grids for area of specimen determination and paricle counting.
  • Translucent
  • Alternative light and dark squares help to reduce eyestrain
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 100499-122
    NE15 Eyepiece Scale
    NE15 Eyepiece Scale
    16 mm
    Log in for availability
    Log in for availability
    Each
    Each - $466.08
    $466.08
    Each
       $466.08
    Supplier Number: 68008-16
  • 100499-124
    NE15 Eyepiece Scale
    NE15 Eyepiece Scale
    19 mm
    Log in for availability
    Log in for availability
    Each
    Each - $466.08
    $466.08
    Each
       $466.08
    Supplier Number: 68008-19
  • 100499-126
    NE15 Eyepiece Scale
    NE15 Eyepiece Scale
    21 mm
    Log in for availability
    Log in for availability
    Each
    Each - $466.08
    $466.08
    Each
       $466.08
    Supplier Number: 68008-21