Specifications
- Description:Static Line II ionizer
- Cat. No.:102681-524
- Supplier no.:SLI
Specifications
About this item
The Static Line II ionizer is an antistatic device which emits negative and positive ions, neutralizing the electrostatic charging in the cryochamber.
The sections no longer stick to the knife edge, or bunch up one on top of the other, but float in a nice ribbon over the knife surface
Applications include: Sucrose protected specimens, cell cultures in elatine/ sucrose, frozen hydrated specimens, dry cut Lowicryls at room temperature, polymers and rubber.
The pickup of the sections can be done easily. Use of the Static Line II ionizer for trimming, sectioning and section collection is recommended.