Lapping and Polishing Fixtures, Electron Microscopy Sciences
About this item
Lapping and polishing fixtures are designed to accurately produce polished parallel, tapered or critically oriented samples with minimal sub-surface damage.
- Tungsten carbide base resists wear and maintains sample planarity
- Wide base ensures stability thereby minimizing sample damage and edge rounding
- Wear ring is replaceable on some fixtures, which extends the life of the fixture
- Micrometer depth control allows precise control over sample thickness
- Precisely crafted stainless steel construction ensures long life and high precision
A series of specialized fixtures has been developed for lapping and polishing samples up to 6" in diameter, TEM samples and oriented crystals. In addition to size and shape, the fixtures also differ in the type of thickness. They are designed to be used either by hand or with a lapping machine for semi-automatic processing (Model 920 above). Sample mounting fixtures are also designed to be used in conjunction with these fixtures to ensure that the sample is mounted parallel to the mounting block and that a thin, uniform layer of wax exists between the mounting block and the sample.
Accessories information: One Vacuum Mounting Block (Series 50153) is required for the Vacuum Mounting Kit and is not included. A custom pattern must be specified by the customer.
2 Options Available Below
Product Details & Documents
Lapping and polishing fixtures are designed to accurately produce polished parallel, tapered or critically oriented samples with minimal sub-surface damage.
- Tungsten carbide base resists wear and maintains sample planarity
- Wide base ensures stability thereby minimizing sample damage and edge rounding
- Wear ring is replaceable on some fixtures, which extends the life of the fixture
- Micrometer depth control allows precise control over sample thickness
- Precisely crafted stainless steel construction ensures long life and high precision
A series of specialized fixtures has been developed for lapping and polishing samples up to 6" in diameter, TEM samples and oriented crystals. In addition to size and shape, the fixtures also differ in the type of thickness. They are designed to be used either by hand or with a lapping machine for semi-automatic processing (Model 920 above). Sample mounting fixtures are also designed to be used in conjunction with these fixtures to ensure that the sample is mounted parallel to the mounting block and that a thin, uniform layer of wax exists between the mounting block and the sample.
Accessories information: One Vacuum Mounting Block (Series 50153) is required for the Vacuum Mounting Kit and is not included. A custom pattern must be specified by the customer.
Product Family Options
Product Information
- AvailabilityPrice
- 101411-658Model 145 Lapping and Polishing FixtureSupplier Number: 50150-45
- 101411-672Model 150 Lapping and Polishing FixtureSupplier Number: 50150-50
Accessories
Product Information
- Catalog #AvailabilityPrice
- Vacuum Mounting Kit
Product Information
- Catalog #AvailabilityPrice
- Micrometer Dual Extension
Product Information
- Catalog #AvailabilityPrice
- Replace Wear Ring Stainless Steel with Tungsten
Product Information
- Catalog #AvailabilityPrice
- Base Extension Adapter