Orders LinkContactUsLinkComponent
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Certificates
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit features FSC-1™ to aid in reducing exposure to formaldehyde gas in the event of a spill. The formalin is transformed into an easily removable semi solid mass. The resulting mass cools rapidly to minimize formaldehyde off gassing. Kits contains 1 – 11oz bottle of FSC-1™ solidifier, 1 pair of vented goggles, a respirator, an open backed gown to protect clothing, a pair of nitrile gloves, 4 scoops and scrapers, 3 wiper pads and 4 thirty gallon bags.
Product Family Options
Product Information
  • Availability
    Price
  • 100503-182
    Formaldehyde Spill Response Refill
    Formaldehyde Spill Response Refill
    Log in for availability
    Log in for availability
    Each
    Each - $385.95
    $385.95
    Each
       $385.95
    Supplier Number: 24141-R
  • 100503-174
    Formaldehyde Spill Response Kt
    Formaldehyde Spill Response Kt
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $466.21
    $466.21
    Kit of 1
       $466.21
    Supplier Number: 24141