Orders LinkContactUsLinkComponent
Paterson® RC Rapid Drying Rack, Electron Microscopy Sciences
Paterson® RC Rapid Drying Rack, Electron Microscopy Sciences
Paterson RC Rapid Drying Rack, 23.6", 23.6"
Catalog # 100492-264
Supplier #:  74839
Paterson® RC Rapid Drying Rack, Electron Microscopy Sciences
Catalog # 100492-264
Paterson® RC Rapid Drying Rack, Electron Microscopy Sciences
Paterson® RC Rapid Drying Rack, Electron Microscopy Sciences
Paterson RC Rapid Drying Rack, 23.6", 23.6"
Price
$160.85
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For air drying up to 20 RC prints in 15 min. Its compact design permits maximum ambient air circulation and prevents trapping of water between the print and the rack. The rack is also suitable for use with fiber-based paper. Print capacity: 5- 11 x 14"; 10-8 x 10"; 20-5 x 7".
Specifications
  • Width:
    23.6"
  • Description:
    Paterson RC Rapid Drying Rack
  • Height:
    23.6"