Pearl® Pipettors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
The lightweight, slender design reduces operator fatigue.
- Entire pipette is autoclavable – no need to recalibrate after autoclaving
- Highly resistant to chemical reagents
- Barrel is designed to minimized dead air space for increased pipetter accuracy