Staining Dish, Electron Microscopy Sciences
Product Details & Documents
Hellendahl type dish for staining or for use as a TLC developing chamber. Holds up to 8 individual 76 x 26 mm (3 x 1") slides, or 16 slides back-to-back.
These dishes are made from soda lime glass. The approximate inside dimensions L x W x D: 76 x 25 x 75 mm.
Specifications
- Description:Staining Hellendahl Dish
- Dimensions:76 x 25 x 75 mm