Specifications
- Description:6-Position Multilap™ Fixture, Model 195
- Cat. No.:100497-040
- Supplier no.:50168-95
Specifications
About this item
In many materials preparation laboratories the need for high specimen throughput and precision made specimen preparation tools is a difficult bottleneck to correct.
- Versatile and precise instrument for performing any precision lapping processes
- Excellent addition to any specimen preparation lab
The Model 195 MultiLapTM is designed to grind and polish multiple specimens with precise control over each individual specimen. The fixture incorporates up to 6 separate pistons which are used for holding the specimen during preparation and controlling the amount of material to be removed. The piston assemblies allow for specimens of up to 1" diameter to be polished in 10-micron increments. The specimen mounts also provide a means for viewing the specimen in transmitted light, enhancing detection of features that require a light transparent specimen.Wear resistant Delrin plastic legs provide support and are very gentle on many types of abrasive films and media.