Orders LinkContactUsLinkComponent
Vacuum Pressure Indicator, Electron Microscopy Sciences
Vacuum Pressure Indicator
  100493-218
 :  71242-01
Vacuum Pressure Indicator, Electron Microscopy Sciences
  100493-218
Vacuum Pressure Indicator, Electron Microscopy Sciences
Vacuum Pressure Indicator, Electron Microscopy Sciences
Vacuum Pressure Indicator
$86.95
Each
This Vacuum-Pressure Indicator fits between the desiccator dome and the desiccator stopcock
Its slips easily onto the tapered outlet of the desiccator dome and into the standard desiccator stopcock without requiring any tools.. The thermo-plastic elastometer collapses when there is negative pressure vacuum in the desiccator and expands under ambient atmospheric pressure.
  • Description:
    Vacuum Pressure Indicator