- Description:Ultrastructure size calculator
- Cat. No.:100502-130
- Supplier no.:74804
A slide-guide-calculator used to determine the real-life size of a structure from a micrograph- in micrometer, nanometer or angstrom units
It computes the real-life dimension/mm at magnifications from 1,000 to 100,000 by introducing a multiplication factor of x2 or x.5. The magnification range can be extended. Waterproof card, printed in English, German and French.