Orders LinkContactUsLinkComponent
MAC Thin Foils Standards for STEM, Electron Microscopy Sciences
MAC Thin Foils Standards for STEM, Electron Microscopy Sciences
Thin Foil Standards for STEM, 25/set
Catalog # 100495-262
Minority or Woman-Owned Business Enterprise
Supplier #:  80082-25
MAC Thin Foils Standards for STEM, Electron Microscopy Sciences
Catalog # 100495-262
Minority or Woman-Owned Business Enterprise
CAS Number:  
MAC Thin Foils Standards for STEM, Electron Microscopy Sciences
MAC Thin Foils Standards for STEM, Electron Microscopy Sciences
Thin Foil Standards for STEM, 25/set
CAS Number:  
Price
$3,746.92
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Selection of 25 high purity thin foils, 0.1 mm thick, 3 mm Ø to fit into TEM grid holders for use in the STEM mode.
  • Polished to a 3 μm diamond finish
Come in a variety of pure metals, including Aluminum, Cadmium, Cobalt, Copper, Gold, Hafnium, Indium, Iron, Magnesium, Molybdenum, Nickel, Niobium, Palladium, Platinum, Rhodium, Silver, Tantalum, Tin, Titanium, Tungsten, Vanadium, Yttrium, Zinc, Zirconium and AISI 316 S/S.
Specifications
Specifications
  • Pack type:
    25/set
  • Description:
    Thin Foil Standards for STEM
  • Cat. No.:
    100495-262