Stereo Viewers, Electron Microscopy Sciences
About this item
A simple device, used for viewing a set of 3-dimensional aerial photos in 3-dimension. It comes with two support legs that are folded flat for storage and are unfolded for the viewing position. Photos are viewed through the two lenses simultaneously with both eyes. Lenses may be adjusted for suitable eye-to-eye width. The coated lenses are 32 mm in diameter.
2X Size: 118 x 56 x 124 mm, Net Weight: 227g
4X Size: 118 x 56 x 82 mm, Net Weight: 216g
1 Options Available Below
Product Details & Documents
A simple device, used for viewing a set of 3-dimensional aerial photos in 3-dimension. It comes with two support legs that are folded flat for storage and are unfolded for the viewing position. Photos are viewed through the two lenses simultaneously with both eyes. Lenses may be adjusted for suitable eye-to-eye width. The coated lenses are 32 mm in diameter.
2X Size: 118 x 56 x 124 mm, Net Weight: 227g
4X Size: 118 x 56 x 82 mm, Net Weight: 216g
Product Family Options
Product Information
- MagnificationAvailabilityPrice
- 100501-592Pocket Mirror Stereo ViewerSupplier Number: 68100
- 100496-268Pocket Mirror Stereo ViewerPocket Mirror Stereo Viewer4×Discontinued-Supplier Number: -
Accessories
Product Information
- Catalog #MaterialGraduationsAvailabilityPrice
- Scale 50, marking area: 50 mm
Product Information
- Catalog #MaterialGraduationsAvailabilityPrice
- Scale 100, marking area: 100 mm
Product Information
- Catalog #MaterialGraduationsAvailabilityPrice
- Scale 200, marking area: 200 mm