ContactUsLinkComponent
Stainless Steel Forceps, with PTFE Tips, Electron Microscopy Sciences
Stainless Steel Forceps, with PTFE Tips, Electron Microscopy Sciences
Catalog # 100501-354
Minority or Woman-Owned Business Enterprise
undefined
Stainless Steel Forceps, with PTFE Tips, Electron Microscopy Sciences
Catalog # 100501-354
Supplier Number:  72978
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Forceps with PTFE Tips, Stainless Steel
  • Cat. No.:
    100501-354
  • Supplier no.:
    72978
Specifications
About this item
Forceps measure 100 mm (4") in overall length and have 38 mm (1 1/16") "PTFE" corrosion resitant tips. Curved shape with a strong fork. Ideal for handling balance weights.
Packaging: Individually blister packed.