Standard Microtome Blade Holder: For use with our disposable blades.
The holders are designed to readily accommodate blades of differing thickness. Such adaptability allows the histologist maximum flexibility especially when difficult tissues are encountered. Durable.
Made from the highest quality stainless steel and the most advanced manufacturing processes, these blade holders are less susceptible to nicks and normal wear and tear.
Convenient, insert, clamp, adjust the angle, and cut. For the first time, the quality and convenience of disposable blades are available for all rotary microtomes and cryostats. Even the popular Miles Tissue Tek™ (adapter required).
Standard Microtome Blade Holder: For use with our disposable blades. 100% stainless steel construction. 2 years warranty. Holder measures: 152×38×10 mm (6×11/2×7/16"). Blade Holder with Extended Handle: Same as Standard except with an extended handle to eliminate knife guard interference. Measures 160 mm (63/8") long. Blade Holder for Low Profile Blades: Same as Standard but designed for use with any low profile microtome blades. Blade Holder for Low Profile with Extended Handle: Same as Low Profile except with an extended handle to eliminate knife guard interference. Blade Holder for High Profile Blade: Same as Standard but designed for use with any High Profile Microtome Blades. Blade Holder for High Profile Blades with Extended Handle: Same as High Profile Blade but comes with an Extended Handle to eliminate knife guard interference. Universal Blade Handle: Patented SHUR/Sharp™ blade holder designed for interchangeable use of Low and High profile blades. Cryostat Adapter: Use when using our Standard blade holder in the Miles TissueTek™ II Cryostat.
100% stainless steel construction. 2 years warranty. Holder measures: 152×38×10 mm (6×11/2×7/16").