Orders LinkContactUsLinkComponent
Slides with Etched Rings, Electron Microscopy Sciences
Slides with Etched Rings, Electron Microscopy Sciences
Slides with Etched Rings, Electron Microscopy Sciences
Certificates
Slides with Etched Rings, Electron Microscopy Sciences
Slides with Etched Rings, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Useful for fluorescent antibody examination.
  • Pre-cleaned
The size of the slide is 3×1", approximately 1 mm thick, with an etched frosted end.
Packaging: Packed 72/box.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 100502-198
    2-etched 15 mm circle slide
    2-etched 15 mm circle slide
    1 g
    Log in for availability
    Log in for availability
    Each
    Each - $727.91
    $727.91
    Each
       $727.91
    Supplier Number: 71879-16
  • 100489-410
    2-etched 10 mm circle slide
    2-etched 10 mm circle slide
    1 g
    Log in for availability
    Log in for availability
    Pack of 144
    Pack of 144 - $83.00
    $83.00
    Pack of 144
       $83.00
    Supplier Number: 71879-10
  • 100489-412
    2-etched 15 mm circle slide
    2-etched 15 mm circle slide
    1 g
    Log in for availability
    Log in for availability
    Pack of 144
    Pack of 144 - $82.04
    $82.04
    Pack of 144
       $82.04
    Supplier Number: 71879-15
  • 100502-196
    2-etched 10 mm circle slide
    2-etched 10 mm circle slide
    1 g
    Log in for availability
    Log in for availability
    Each
    Each - $720.63
    $720.63
    Each
       $720.63
    Supplier Number: 71879-11