SaniZide™ Germicidal Solution, Electron Microscopy Sciences
Ready to use, hospital grade, hard surface disinfectant deodorizer.
- Quatrernary ammonium, alcohol free formulation
- Non-flammable and non-corrosive
- SaniZide Plus™ safe to ship
Choosing SaniZide Plus™ helps you to comply with the OSHA Bloodborne Pathogens Standard, which requires the use of an “appropriate disinfectant” that is tuberculocidal and virocidal against HIV-1 and HBV.
Safe for use on: glass, porcelain, ceramic, metal, polyethylene, polypropylene, vinyl, polyester, rubber, bakelite, and many more.
: Packaging in a convenient spray dispenser bottle, either by pump or trigger.