ContactUsLinkComponent
Sample Vials In Mini-File®; Clear, Electron Microscopy Sciences
Sample Vials In Mini-File®; Clear, Electron Microscopy Sciences
Catalog # 100490-068
Minority or Woman-Owned Business Enterprise
undefined
Sample Vials In Mini-File®; Clear, Electron Microscopy Sciences
Catalog # 100490-068
Supplier Number:  60996-02
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Cap included:
    Yes
  • Capacity:
    2.00 ml
  • Description:
    Sample vials in Mini-File
  • Cat. No.:
    100490-068
  • Supplier no.:
    60996-02
Specifications
About this item
These vials are made from Wheaton-33R a low extractable borosilicate glass.
  • Vials and caps can be autoclaved
Delivery: The vials are supplied with a white (styrene-butadiene) rubber lined cap.
Packaging: Comes ten vials packed in a clean plastic thermoformed Mini-File and offers convenience in transporting, displaying and storing.