Orders LinkContactUsLinkComponent
Rubber Cement, Electron Microscopy Sciences
Rubber Cement, Electron Microscopy Sciences
Rubber Cement
Catalog # 100491-938
Supplier #:  72170
Rubber Cement, Electron Microscopy Sciences
Catalog # 100491-938
CAS Number:  
Rubber Cement, Electron Microscopy Sciences
Rubber Cement, Electron Microscopy Sciences
Rubber Cement
CAS Number:  
Price
$12.53
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Excellent adhesive for all paper pasting and mountings of drawings and photographs. It is also used to adhere specimens to SEM Stubs. Any excess cured cement can be easily removed. 4 oz. bottle comes with a brush in the cap.
Certifications: Certificates of analysis are available.
Documents
Certificates
Search by Lot Number
Specifications
Specifications
  • Description:
    Rubber Cement