Orders LinkContactUsLinkComponent
MAC Rare Earth Set for TEM, Electron Microscopy Sciences
MAC Rare Earth Set for TEM, Electron Microscopy Sciences
Rare Earth Set, 14/set
Catalog # 100495-264
Supplier #:  80086-14
MAC Rare Earth Set for TEM, Electron Microscopy Sciences
Catalog # 100495-264
CAS Number:  
MAC Rare Earth Set for TEM, Electron Microscopy Sciences
Rare Earth Set, 14/set
CAS Number:  
Price
$1,739.76
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This set consists of 14 rare earth compounds dispersed onto holey carbon films (3.05 mm, 400# copper grid).
The compunds are LaF3, CeAl2, PrF3, NdF3, SmF3, EuF3, GdF3, TbF3, DyF3, HoF3, ErF3, TmF3, YbF3, and LuF3.
Specifications
Specifications
  • Pack type:
    14/set
  • Description:
    Rare Earth Set