ContactUsLinkComponent
Peca Techni-Valve™, Electron Microscopy Sciences
Peca Techni-Valve™, Electron Microscopy Sciences
Catalog # 100502-170
Minority or Woman-Owned Business Enterprise
undefined
Peca Techni-Valve™, Electron Microscopy Sciences
Catalog # 100502-170
Supplier Number:  70849-10
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Peca Techni-Valve™
  • Cat. No.:
    100502-170
  • Supplier no.:
    70849-10
Specifications
About this item
Peca Techni-Valve™ for use with Dust-Pro™ Refill cans.
  • Smooth bell housing and no mounting clip is required
  • Patented bell housing fits the contour of your hand
It is made from solid brass chrome plated is professionally designed with a long nozzle (90 mm (31/2")). The valve can be removed at any time without the loss of pressure.