ContactUsLinkComponent
Print Trimmer, Electron Microscopy Sciences
Print Trimmer, Electron Microscopy Sciences
Catalog # 100501-334
Minority or Woman-Owned Business Enterprise
undefined
Print Trimmer, Electron Microscopy Sciences
Catalog # 100501-334
Supplier Number:  70860
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Print Trimmer
  • Dimensions:
    51/2×171/2"
  • Cat. No.:
    100501-334
  • Supplier no.:
    70860
Specifications
About this item
This print trimmer has super sharp cutting edge, using a standard single edge razor blade.
  • Handles prints up to 14"
  • Light weight
This provides unobstructed view of the cutting process. Designed with safe, fully enclosed retractable blade.