Specifications
- Description:Photomicrograph scale marker
- Cat. No.:100492-238
- Supplier no.:74802
Specifications
About this item
A slide-ruler used to apply scale marks to photomicrographs.
- Made from a clear flexible plastic and it is protected by an enclosed pouch
- Instructions are included
- Printed in English, German, and French
There are four scale lines for producing scale marks of 10 microns, 1 micron, 0.1 micron (100 nm) and 10 nm. The scale marker is also used in measuring the real life size of an object in a micrograph.