Specifications
- Description:Petri Dish Analyslide
- Cat. No.:100488-402
- Supplier no.:70650
Specifications
About this item
The analyslide is like a petri dish on a slide. Use it as an examination and storage container for 47 mm filters.
- Rectangular base fits any microscope stage
- Inner cover ring offers airtight security for the samples and it does not interfere with viewing
- Ideal for drying coated grids and absorbing water from grids that are supporting sections