Order Entry
United States
ContactUsLinkComponent
 
Paterson® RS Print Squeegee, Electron Microscopy Sciences
  100492-260
Minority or Woman-Owned Business Enterprise
undefined
Paterson® RS Print Squeegee, Electron Microscopy Sciences
  100492-260
 :  74823
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    Paterson RS Print Squeegee
  • Environmentally Preferable:
  • Cat. No.:
    100492-260
  • Supplier no.:
    74823

 

 

Extra soft rubber blades and a unique floating-action lower blade prevents any damage to the print surface.

  • Suitable for both RS and fiber-based papers
  • The print squeegee handles prints up to 40×50 cm (16× 20")