Orders LinkContactUsLinkComponent
Paterson® Auto Print Washers, Electron Microscopy Sciences
Paterson® Auto Print Washers, Electron Microscopy Sciences
Paterson® Auto Print Washers, Electron Microscopy Sciences
Certificates
Paterson® Auto Print Washers, Electron Microscopy Sciences
Paterson® Auto Print Washers, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Paterson® Standard and Major Print Washers.
  • The Major holds 12 (11 x 14") or 24 (5 x 7") prints
  • The Standard holds up to 12 (8 x 10") or 24 (5 x 7") prints*
  • All prints remain separate and water freely circulates
Paterson® Standard print washer is perfect for washing both RC and fiber-based papers. To use, prints are loaded into the cradle which is then lowered into the tank. Faucet pressure gently rocks the print cradle as water continuously exchanges.
Paterson® Major print washer is similar to the standard one, but with a larger capacity and a built-in siphon for convenient emptying after use.
* Note: For our international users: Standard and Major Print Washers will in fact accept prints to 24 x 30 cm (10 x 12") and 30 x 28 cm (12 x 15"), respectively.
Product Family Options
Product Information
  • Availability
    Price
  • 100494-000
    Paterson Print Washer
    Paterson Print Washer
    Log in for availability
    Log in for availability
    Each
    Each - $405.05
    $405.05
    Each
       $405.05
    Supplier Number: 74353-10
  • 100494-002
    Paterson Major Print Washer
    Paterson Major Print Washer
    Log in for availability
    Log in for availability
    Each
    Each - $811.30
    $811.30
    Each
       $811.30
    Supplier Number: 74353-20