Orders LinkContactUsLinkComponent
Numbered Grid Storage Box, 100 Capacity, Electron Microscopy Sciences
Numbered Grid Storage Box, 100 Capacity, Electron Microscopy Sciences
Numbered Grid Storage Box, 100 Capacity, Electron Microscopy Sciences
Certificates
Numbered Grid Storage Box, 100 Capacity, Electron Microscopy Sciences
Numbered Grid Storage Box, 100 Capacity, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This is the standard 100 capacity grid storage box with a unique number printed on the face and on one end.
  • Eliminates the placement of the specimen grid in the wrong box
  • Easy retrieval of grid box from storage
  • Complete with grid recording card
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 100491-740
    Numbered Grid Storage Box
    Numbered Grid Storage Box
    1/Pack
    Log in for availability
    Log in for availability
    Each
    Each - $41.93
    $41.93
    Each
       $41.93
    Supplier Number: 71137
  • 100496-316
    Numbered Grid Storage Box
    Numbered Grid Storage Box
    10/Lot
    Log in for availability
    Log in for availability
    Each
    Each - $358.91
    $358.91
    Each
       $358.91
    Supplier Number: 71138
  • 100496-318
    Numbered Grid Storage Box
    Numbered Grid Storage Box
    100/Lot
    Log in for availability
    Log in for availability
    Each
    Each - $3,290.57
    $3,290.57
    Each
       $3,290.57
    Supplier Number: 71139