Specifications
- Description:Nail Polish
- Size:0.5 oz
- Cat. No.:100491-940
- Supplier no.:72180
Specifications
About this item
Nail polish is used for sealing the boats made from tape to the glass knives.
Nail polish is used for sealing the boats made from tape to the glass knives.