Tripod Polisher® TEM/SEM Sample Preparation, 590, Electron Microscopy Sciences
About this item
A Tripod Polisher® was designed to prepare accurately micro sizes of TEM and SEM samples.
- Limited ion milling
- No strong chemicals are used to prepare the specimen
- Reduce preferential thinning, radiation damage, and heating of the sample
Used to prepare both plan-view and cross-sections from a variety of sample materials, such as ceramics, composites, metals and, geological specimens. The same sample can be studied with both SEM and TEM and hence reduces sample preparation.
23 Options Available Below
Product Details & Documents
A Tripod Polisher® was designed to prepare accurately micro sizes of TEM and SEM samples.
- Limited ion milling
- No strong chemicals are used to prepare the specimen
- Reduce preferential thinning, radiation damage, and heating of the sample
Used to prepare both plan-view and cross-sections from a variety of sample materials, such as ceramics, composites, metals and, geological specimens. The same sample can be studied with both SEM and TEM and hence reduces sample preparation.
Product Family Options
Product Information
- AvailabilityPrice
- 100491-340Starter Kit for Tripod Polisher®Supplier Number: 50118-05
- 100496-836Micrometer BaseSupplier Number: 50116-05
- 100496-838Precision Locking MicrometerSupplier Number: 50116-10
- 100496-840Plain "L" BracketSupplier Number: 50116-20
- 100496-842Screw Clamp "L" BracketSupplier Number: 50116-25
- 100496-844Clamping "L" BracketSupplier Number: 50116-30
- 100496-846X-Section L-BracketSupplier Number: 50116-31
- 100496-848Sample HolderSupplier Number: 50116-35
- 100496-850Planarizing ToolSupplier Number: 50116-40
- 100496-854Heater BlockSupplier Number: 50116-50
- 100496-856Pedestal for Model 590Supplier Number: 50116-55
- 100496-858Pyrex InsertSupplier Number: 50116-60
- 100496-860Wedge Polishing MountSupplier Number: 50116-61
- 100496-862Wedge Polishing ClampSupplier Number: 50116-62
- 100496-864Pyrex Wedge Polishing RodSupplier Number: 50116-63
- 100496-866Pyrex Wedge Polishing StubSupplier Number: 50116-64
- 100496-868Glass Leveling SlideSupplier Number: 50116-70
- 100496-870Tool KitTool KitDiscontinued-Supplier Number: -
- 100502-428Derlin Micrometer FeeSupplier Number: 50116-15
- 100502-430Parallel Polishing Mount, Stainless SteelSupplier Number: 50116-36
- 100503-044Stub AssemblySupplier Number: 50116-65
- 101411-584Tripod Polisher® 590, Kit for TemSupplier Number: 50115-10
- 101411-586Tripod Polisher® 590, Kit for SEMSupplier Number: 50115-20
- 101411-588Tripod Polisher® 590, Kit for TEM and SEMSupplier Number: 50115-30