ContactUsLinkComponent
Mini Vacuum Desiccator, Electron Microscopy Sciences
  100493-228
Minority or Woman-Owned Business Enterprise
undefined
Mini Vacuum Desiccator, Electron Microscopy Sciences
  100493-228
 :  71246-01
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Mini Vacuum Desiccator
  • Cat. No.:
    100493-228
  • Supplier no.:
    71246-01
The top part of the desiccator is clear, molded from polycarbonate and see-through from any angle. A polyethylene stopcock with a "PTFE" plug allows for gentle releasing of the vacuum. A peripheral flange around the polypropylene bottom section fits into and snap locks therefore sealing the two halves against a "O" ring. Vacuum retention rating is 740 mm Hg (29"). Inside dimensions are 102mm (4") diameter x 59mm (2 7/16")high. The base is 175mm (6 7/8") diameter.
  • Ideal for Small Samples
  • Degassing Embedding mixtures