ContactUsLinkComponent
Microtome Oil, Electron Microscopy Sciences
Microtome Oil, Electron Microscopy Sciences
Catalog # 100504-370
Minority or Woman-Owned Business Enterprise
undefined
Microtome Oil, Electron Microscopy Sciences
Catalog # 100504-370
Supplier Number:  60744
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Microtome Oil
  • Size:
    100 ml
  • Cat. No.:
    100504-370
  • Supplier no.:
    60744
Specifications
About this item
A chemically pure, high-grade lubricant used in precision instruments. Should be used periodically to lubricate cross-rollerbearings for microtomes.