Water/Ice Bath Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Rust-proof anodized aluminum rack – designed to be used in water/ice baths.
- Removable cover keeps tubes closed while boiling
- Handle for easy removal from water baths
- Will not rust