Macro-Cassette, Electron Microscopy Sciences
Product Details & Documents
Made from Acetal-Polymer. Comes complete with a one piece integral lid . It is specially designed to hold large specimens during the embedding process, as well as in a storage cabinet. Inside measurements L x W x D: 29 x 25 x 11 mm.
Specifications
- Description:Macro Cassette, White