Low Intensity UV Lamp, Electron Microscopy Sciences
About this item
This 6-watt UV lamp meets most laboratory requirements; such as, bacterial identification, fluorchemistry, specimen staining, titration processes, photochemical reactions, aflatoxin detection, and curing UV cements.
Constructed from Cyolac® plastic, with a longwave emission of 365 nm and an intensity at 6" (15.2 cm) of 750 µm/cm². The bulb has a rated life of 6000 hours. It can be used as a hand-held light or it may br mounted on a optional stand. Powered by 110 V/60 Hz or 220 V/50 Hz.
4 Options Available Below
Product Details & Documents
This 6-watt UV lamp meets most laboratory requirements; such as, bacterial identification, fluorchemistry, specimen staining, titration processes, photochemical reactions, aflatoxin detection, and curing UV cements.
Constructed from Cyolac® plastic, with a longwave emission of 365 nm and an intensity at 6" (15.2 cm) of 750 µm/cm². The bulb has a rated life of 6000 hours. It can be used as a hand-held light or it may br mounted on a optional stand. Powered by 110 V/60 Hz or 220 V/50 Hz.
Product Family Options
Product Information
- VoltageAvailabilityPrice
- 100491-958Low-Intensity UV LampSupplier Number: 72422Specifications:Minority or Woman-Owned Business Enterprise
More
Specifications
Cat. No.100491-958Minority or Woman-Owned Business Enterprise - 100491-964Low-Intensity BulbSupplier Number: 72425Specifications:Minority or Woman-Owned Business Enterprise
More
Specifications
Cat. No.100491-964Minority or Woman-Owned Business Enterprise - 100491-960Low-Intensity UV LampSupplier Number: 72423Specifications:Minority or Woman-Owned Business Enterprise
More
Specifications
Cat. No.100491-960Minority or Woman-Owned Business Enterprise - 100491-962Low-Intensity Uv Lamp StandSupplier Number: 72424Specifications:Minority or Woman-Owned Business Enterprise
More
Specifications
Cat. No.100491-962Minority or Woman-Owned Business Enterprise