Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lab Spoon, Electron Microscopy Sciences

Lab Spoon, Electron Microscopy Sciences

Lab spoon, 229 mm
Catalog # 100492-066
Supplier #:  72987
Lab Spoon, Electron Microscopy Sciences
Catalog # 100492-066
Lab Spoon, Electron Microscopy Sciences
Lab Spoon, Electron Microscopy Sciences

Lab spoon, 229 mm

Price
$93.07
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Spoon has 4 mm (⁵/₃₂") Ø stainless steel rod, flattened at one end to give a 8×51 mm blade. The other end has a 13×25 mm spoon.

The blade as well as the spoon is coated with 'PTFE'. Overall length 229 mm (9").

Specifications
  • Description:
    Lab spoon
  • Length:
    229 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.