ContactUsLinkComponent
KOMET™ Spinbar® - Stirring Bar, Electron Microscopy Sciences
KOMET™ Spinbar® - Stirring Bar, Electron Microscopy Sciences
Catalog # 100498-846
Minority or Woman-Owned Business Enterprise
undefined
KOMET™ Spinbar® - Stirring Bar, Electron Microscopy Sciences
Catalog # 100498-846
Supplier Number:  64517-01
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Coating:
    PTFE
  • Description:
    Komet Spin Bar
  • Cat. No.:
    100498-846
  • Supplier no.:
    64517-01
Specifications
About this item
The 'PTFE' Komet™ Spinbar® Magnetic Stirring Bar is made from a high-energetic magnetic material.
  • Improved efficiency
  • The Samarium-Cobalt magnet is encapsulated in 'PTFE'
  • The risk of demagnetization caused by external magnetic fields is completely eliminated
Combined with its octagonal shape, produce exceptional power although its length is only 50 mm (1.969") with a 21 mm (0.82") diameter. Its eight edges generate strong turbulence so liquids are thoroughly mixed and its stability is not impaired in vessels with curved bottoms. Komet™ transmits torque loads two to three times larger than those of conventional stirring bars of this length significantly improving efficiency.