About this item
To convert your Model 4.0 Secador™ Autodesiccator Cabinets or Secador™ Desiccator Cabinets from the Horizontal configuration to the Vertical, or from Vertical configuration to Horizontal, all you need is a new set of shelves.