SECADOR™ 1.0 Desiccator Cabinet, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
- Built-in hygrometer - Quick check on RH levels.
- Transparent for easy viewing of contents.
- Superior chemical resistance - Resists staining.
- Patented sealed construction - Airtight.
- Stackable - Small footprint.
Accessories information: Carrying case with two latches and handle available for portable convenience.