Orders LinkContactUsLinkComponent
Spill Control Tray, Electron Microscopy Sciences
Spill Control Tray, Electron Microscopy Sciences
Spill Control Tray
Catalog # 100496-222
Supplier #:  64448
Spill Control Tray, Electron Microscopy Sciences
Catalog # 100496-222
Spill Control Tray, Electron Microscopy Sciences
Spill Control Tray, Electron Microscopy Sciences
Spill Control Tray
Price
$151.35
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The tray is made from vacuum formed polystyrene 12.7 cm (1/2") deep with an external flange.The overall dimensions are 58 x 69 cm (23 x 27"). The tray accommodates our Benchtop mat. These trays are very useful in the biology or chemistry labs.
Specifications
  • Description:
    Spill Control Tray